Merck
  • Complex metabolic interactions between benzo(a)pyrene and tributyltin in presence of dichlorodiphenyltrichloroethane in South American catfish Rhamdia quelen.
Complex metabolic interactions between benzo(a)pyrene and tributyltin in presence of dichlorodiphenyltrichloroethane in South American catfish Rhamdia quelen.
Ecotoxicology and environmental safety (2013-07-23)
Heloísa H P Oliveira, Mathieu Babin, Juan Ramon Esquivel Garcia, Francisco Filipak Neto, Marco A F Randi, Ciro A Oliveira Ribeiro, Émilien Pelletier
ZUSAMMENFASSUNG
In an attempt to explore complex metabolic interactions between toxicants present in polluted freshwater, hepatic metabolism of benzo(a)pyrene (BaP) and tributyltin (TBT) in fish was investigated when these compounds were administrated alone, mixed together and along with dichlorodiphenyltrichloroethane (DDT). Ten Rhamdia quelen per group were treated with a single intra-peritoneal (IP) dose (5-day experiment) or three successive doses (15-day experiment) either containing BaP (0.3; 3 or 30mgkg(-1)) or TBT (0.03; 0.3 or 3mgkg(-1)) or a combination of BaP+TBT, BaP+DDT, TBT+DDT and BaP+TBT+DDT under their respective lower doses, with DDT dose kept at 0.03mgkg(-1). Tetrahydroxy-benzo(a)pyrene (BaP-tetrol-I), and dibutyltin (DBT) and monobutyltin (MBT) were analyzed to assess BaP and TBT hepatic metabolism, respectively. A significant difference in BaP-tetrol-I concentration was observed in liver and bile between the lowest and the highest doses of BaP in both 5 and 15-day experiments. In the 15-day experiment, the presence of TBT with BaP reduced the amount of BaP-tetrol-I in bile compared to the BaP alone. The time of exposure and the number of doses affected BaP-tetrol-I concentration in the bile of fish exposed to BaP 0.3mgkg(-1) and BaP+DDT. TBT and its metabolites concentrations showed a dose-dependent increase in the liver in both experiments and in the bile in the 5-day experiment. TBT at its lowest dose was completely metabolized into DBT and MBT in the liver in the 15-day experiment. No TBT metabolites were detected in the bile of fish exposed to the mixtures in the 5-day experiment, except for a small MBT amount found in BaP+TBT+DDT. This study strengthens the hypothesis of a metabolic interaction between BaP and TBT in fish and suggests DDT as an important third player when present in the mixture.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzo[a]pyren, ≥96% (HPLC)
Sigma-Aldrich
Tributylzinnhydrid, contains 0.05% BHT as stabilizer, 97%
Supelco
Benzo[a]pyren, analytical standard, for environmental analysis
Supelco
Benzo[a]pyren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
4,4′-DDT, PESTANAL®, analytical standard
Supelco
Benzo[a]pyren -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
4,4′-DDT -Lösung, certified reference material, 5000 μg/mL in methanol
Supelco
Benzo[a]pyren -Lösung, 100 μg/mL in cyclohexane, analytical standard