Merck
  • Co3O4-carbon nanotube heterostructures with bead-on-string architecture for enhanced lithium storage performance.
Co3O4-carbon nanotube heterostructures with bead-on-string architecture for enhanced lithium storage performance.
Nanoscale (2013-07-24)
Minwei Xu, Fei Wang, Yin Zhang, Sen Yang, Mingshu Zhao, Xiaoping Song
ZUSAMMENFASSUNG
In this report, alkylcarboxyl group-decorated carbon nanotubes (CNTs) with clustered functionalization patterns are achieved based on a modified Birch reduction in liquid ammonia. By using these functional CNTs (f-CNTs), a new type of Co3O4-CNT heterostructure is prepared via a simple hydrothermal method. SEM and TEM analyses reveal that the as-synthesized Co3O4-CNT heterostructures exhibit bead-on-string architecture, in which the Co3O4 spheres are threaded with CNTs. A possible growth mechanism is proposed to explain the formation of these Co3O4-CNT heterostructures. The electrochemical properties of the Co3O4-CNT heterostructures as anode materials for lithium ion batteries are investigated. The Co3O4-CNT heterostructures display high electrochemical activity, good cycle stability and improved rate performance. Such a large improvement of the electrochemical performance can be related to the robust necklace-like architectures which possess properties such as high mechanical stability, excellent electric conductivity and good strain accommodation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis