Merck
One-pot squaric acid diester mediated aqueous protein conjugation.
Chemical communications (Cambridge, England) (2013-07-26)
Frederik Wurm, Tobias Steinbach, Harm-Anton Klok
ZUSAMMENFASSUNG
A water-soluble squaric acid dialkyl diester derivative is presented, which enables one-pot, two-step amine-selective protein conjugation reactions with (functional) amines in water. This reagent not only allows all-aqueous protein modifications, but also tolerates e.g. hydroxyl groups and can also be used for the modification of proteins with water-insoluble amines.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Dihydroxy-3-cyclobuten-1,2-dion, ≥99.0% (HPLC)