Merck
  • CORR Insights®: Staphylococcus aureus colonization among arthroplasty patients previously treated by decolonization protocol: a pilot study.
CORR Insights®: Staphylococcus aureus colonization among arthroplasty patients previously treated by decolonization protocol: a pilot study.
Clinical orthopaedics and related research (2013-07-26)
Javad Parvizi
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mupirocin, 920-1020 μg/mg (HPLC), crystalline
Sigma-Aldrich
Lithium-Mupirocin Lithiumsalz, ≥95.0% (HPLC)
Millipore
Lithium-Mupirocin-Supplement, suitable for microbiology,
Selectively inhibits the growth of lactic acid bacteria