Merck
The Akt substrate Girdin is a regulator of insulin signaling in myoblast cells.
Biochimica et biophysica acta (2013-07-28)
Angelika Hartung, Anna-Maria Ordelheide, Harald Staiger, Martina Melzer, Hans-Ulrich Häring, Reiner Lammers
ZUSAMMENFASSUNG
Akt kinases are important mediators of the insulin signal, and some Akt substrates are directly involved in glucose homeostasis. Recently, Girdin has been described as an Akt substrate that is expressed ubiquitously in mammals. Cells overexpressing Girdin show an enhanced Akt activity. However, not much is known about Girdin's role in insulin signaling. We therefore analyzed the role of Girdin in primary human myotubes and found a correlation between Girdin expression and insulin sensitivity of the muscle biopsy donors, as measured by a hyperinsulinemic-euglycemic clamp. To understand this finding on a cellular level, we then investigated the function of Girdin in C2C12 mouse myoblasts. Girdin knock-down reduced Akt and insulin receptor substrate-1 phosphorylation. In contrast, stable overexpression of Girdin in C2C12 cells strikingly increased insulin sensitivity through a massive upregulation of the insulin receptor and enhanced tyrosine phosphorylation of insulin receptor substrate-1. Furthermore, Akt and c-Abl kinases were constitutively activated. To investigate medium-term insulin responses we measured glucose incorporation into glycogen. The Girdin overexpressing cells showed a high basal glycogen synthesis that peaked already at 1nM insulin. Taken together, we characterized Girdin as a new and major regulator of the insulin signal in myoblasts and skeletal muscle.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycogen aus Austern, ≥75% dry basis
Sigma-Aldrich
Glycogen aus Rinderleber, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glykogen aus Muscheln, Mytilus-Gattung aus Mytilus edulis (Miesmuschel), for DNA precipitations
Sigma-Aldrich
Glycogen aus Kaninchenleber, ≥85% dry basis (enzymatic)
Sigma-Aldrich
Glykogen aus Muscheln, Mytilus-Gattung aus Mytilus edulis (Miesmuschel), ≥85% anhydrous basis (enzymatic)
Sigma-Aldrich
Anti-IRS1-Antikörper, Upstate®, from rabbit