Merck
Update on neuropathic pain treatment: ion channel blockers and gabapentinoids.
Current pain and headache reports (2013-07-28)
Lucy Chen, Jianren Mao
ZUSAMMENFASSUNG
Neuropathic pain is a debilitating chronic pain condition, which remains difficult to treat. The current mainstays of treatment include physical therapy, interventional procedures and medications. Among medications, ion channel blockers and gabapentinoids are the 2 classes of drugs commonly used to treat neuropathic pain. It has been suggested that these medications may be useful to treat a variety of neuropathic pain conditions. This article provides several updates on the utility of both ion channel blockers and gabapentinoids for the treatment of neuropathic pain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Gabapentin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Gabapentin -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Gabapentin, solid