Merck
  • Laminin production and basement membrane deposition by mesenchymal stem cells upon adipogenic differentiation.
Laminin production and basement membrane deposition by mesenchymal stem cells upon adipogenic differentiation.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2013-08-01)
Ariel Noro, Tarvo Sillat, Ismo Virtanen, Sulev Ingerpuu, Nils Bäck, Yrjö T Konttinen, Matti Korhonen
ZUSAMMENFASSUNG
The aim was to study laminin (LM) synthesis, integration, and deposition into the basement membrane (BM) during adipogenesis. Human bone marrow-derived mesenchymal stromal cells (MSCs) were induced along the adipogenic lineage. LM chain mRNA and protein levels were followed using quantitative real-time polymerase chain reaction (qRT-PCR), immunofluorescence (IF) staining, transmission electron microscopy (TEM), and immunoprecipitation. MSCs produced low levels of LM mRNAs but were not surrounded by BM in IF and TEM imaging. LM-α4, LM-β1, and LM-γ1 mRNAs increased during adipogenesis 3.9-, 5.8-, and 2.8-fold by day 28. LM-411 was immunoprecipitated from the ECM of the differentiated cells. Immunostaining suggested deposition of LM-411 and some LM-421. BM build-up was probably organized in part by integrin (Int) α6β1. At day 28, TEM images revealed BM-like structures around fat droplet-containing cells. The first signs of BM formation and Int α6β1 were seen using IF imaging at day 14. Laminin-411 and Int α6β1 were expressed in vivo in mature human subcutaneous fat tissue. Undifferentiated human MSCs did not organize LM subunits into BM, whereas LM-411 and some LM-421 are precipitated in the BM around adipocytes. This is the first demonstration of LM-411 precipitation during hMSC adipogenesis around adipocytes as a structural scaffold and Int-regulated signaling element.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ölrot O -Lösung, 0.5% in isopropanol
Sigma-Aldrich
Ölrot O, certified by the BSC
Sigma-Aldrich
Ölrot O -Lösung, 0.5% in propylene glycol
Supelco
Ölrot O, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Anti-Integrin α6 Antibody, clone NKI-GoH3, clone NKI-GoH3, Chemicon®, from rat