Merck
Locating Pb2+ and Zn2+ in zinc finger-like peptides using mass spectrometry.
Journal of the American Society for Mass Spectrometry (2013-08-03)
Laura Banu, Voislav Blagojevic, Diethard K Bohme
ZUSAMMENFASSUNG
The binding preferences of Pb(2+)and Zn(2+) in doubly charged complexes with zinc finger-like 12-residue peptides (Pep), [Mn(Pep-2(n-1)H)](2+) have been explored using tandem mass spectrometry. The peptides were synthesized strategically by blocking the N-terminus with an acetyl group and with four cysteine and/or histidine residues in positions 2, 5, 8, and 11, arranged in different motifs: CCHH, CHCH, and CCCC. The MS(2) spectra of the Pb(2+) and Zn(2+) complexes show multiple losses of water and a single methane loss and these provide a sensitive method for locating the metal dication and so elucidating its coordination. The elimination of a methane molecule indicated the position of the metal at the Cys2 residue. Whereas lead was observed to preferentially bind to cysteine residues, zinc was found to primarily bind to histidine residues and secondarily to cysteine residues. Preferential binding of lead to cysteine is preserved in the complexes with more than one Pb(2+). Key to the mechanism of the loss of water and methane is the metal dication withdrawing electrons from the proximal amidic nitrogen. This acidic nitrogen loses its hydrogen to an amidic oxygen situated four atoms away leading to formation of a five-member ring and the elimination of water.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Histidin, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidin, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidin
Supelco
L-Histidin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Histidin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland