Merck
Development of a new tissue-equivalent material applied to optimizing surgical accuracy.
Materials science & engineering. C, Materials for biological applications (2013-08-06)
Shan Jiang, Zhiliang Su, Xingji Wang, Sha Liu, Yan Yu
ZUSAMMENFASSUNG
The precision of orientation to target placement during invasive therapy is mainly influenced by tool-tissue interaction. In this study, we aim to investigate a transparent Poly (vinyl alcohol) (PVA) hydrogel as tissue-equivalent material which is used in accurate surgical insertion research. The PVA hydrogel with specified formula was prepared by means of physical and chemical crosslink. The effects of chemical composition and synthesis technique on the biomechanical property, density and micro-structure morphology of PVA materials have been investigated in detail. It can be concluded that when PVA concentration is 8 g/dl, the NaCl concentration is 4 wt.%, with mix water/DMSO solvent, prepared under 7 freeze/thaw cycles, the material has the most similar properties with kidney tissue. Experimental results demonstrate that this tissue-equivalent material could be used in the ex vivo insertion accuracy test for robot-assisted percutaneous intervention and surgical training in minimally invasive surgery (MIS).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(vinylalkohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 8,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 150,000 (average), 99+% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinylalkohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 85,000-124,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinylalkohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(vinylalkohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinylalkohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Poly(vinylalkohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Kuraray Poval® 56-98, Mw ~195,000
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylenglycol)-dimethacrylat, average Mn 20,000, contains MEHQ as inhibitor