Merck
Lower, faster, better: the delusions of surrogate markers in hypertension.
International journal of clinical practice (2013-08-21)
A S Wierzbicki, A Ferro
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Amlodipinbesilat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Amlodipinbesilat, ≥98% (HPLC)
Amlodipinbesilat, European Pharmacopoeia (EP) Reference Standard
Amlodipin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard