Merck
  • One-pot formation of 1,3,4-oxadiazol-2(3H)-ones and dibenzo[c,e]azepines by concomitant cathodic reduction of diazonium salts and phenanthrenequinones.
One-pot formation of 1,3,4-oxadiazol-2(3H)-ones and dibenzo[c,e]azepines by concomitant cathodic reduction of diazonium salts and phenanthrenequinones.
The Journal of organic chemistry (2013-08-21)
Belen Batanero, Fructuoso Barba, Avelino Martin
ZUSAMMENFASSUNG
The one-pot concomitant electrochemical reduction of phenanthrenequinones (1, 2) and arenediazonium salts (3a-f) led to the formation of 1,3,4-oxadiazol-2(3H)-ones (4a-f, 5a) and dibenzo[c,e]azepines (6a-f) when N-methylformamide was used as the solvent. A new pathway, different from those previously described with other aprotic solvents, is proposed. The experimental data support a radical mechanism for the electrochemical process followed by an internal rearrangement to give the products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Clozapin
Sigma-Aldrich
9,10-Phenanthrenchinon, 95%
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
9,10-Phenanthrenchinon, ≥99%
Clozapin, European Pharmacopoeia (EP) Reference Standard
Clozapin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard