Merck
  • Comparison of large scale purification processes of naproxen enantiomers by chromatography using methanol-water and methanol-supercritical carbon dioxide mobile phases.
Comparison of large scale purification processes of naproxen enantiomers by chromatography using methanol-water and methanol-supercritical carbon dioxide mobile phases.
Journal of chromatography. A (2013-08-21)
Fahimeh Kamarei, Péter Vajda, Georges Guiochon
ZUSAMMENFASSUNG
This paper compares two methods used for the preparative purification of a mixture of (S)-, and (R)-naproxen on a Whelk-O1 column, using either high performance liquid chromatography or supercritical fluid chromatography. The adsorption properties of both enantiomers were measured by frontal analysis, using methanol-water and methanol-supercritical carbon dioxide mixtures as the mobile phases. The measured adsorption data were modeled, providing the adsorption isotherms and their parameters, which were derived from the nonlinear fit of the isotherm models to the experimental data points. The model used was a Bi-Langmuir isotherm, similar to the model used in many enantiomeric separations. These isotherms were used to calculate the elution profiles of overloaded elution bands, assuming competitive Bi-Langmuir behavior of the two enantiomers. The analysis of these profiles provides the basis for a comparison between supercritical fluid chromatographic and high performance liquid chromatographic preparative scale separations. It permits an illustration of the advantages and disadvantages of these methods and a discussion of their potential performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthalinessigsäure, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Naproxen -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard