Merck
  • Ribonucleases as a host-defence family: evidence of evolutionarily conserved antimicrobial activity at the N-terminus.
Ribonucleases as a host-defence family: evidence of evolutionarily conserved antimicrobial activity at the N-terminus.
The Biochemical journal (2013-08-22)
Marc Torrent, David Pulido, Javier Valle, M Victòria Nogués, David Andreu, Ester Boix
ZUSAMMENFASSUNG
Vertebrate secreted RNases (ribonucleases) are small proteins that play important roles in RNA metabolism, angiogenesis or host defence. In the present study we describe the antimicrobial properties of the N-terminal domain of the hcRNases (human canonical RNases) and show that their antimicrobial activity is well conserved among their lineage. Furthermore, all domains display a similar antimicrobial mechanism, characterized by bacteria agglutination followed by membrane permeabilization. The results of the present study show that, for all antimicrobial hcRNases, (i) activity is retained at the N-terminus and (ii) the antimicrobial mechanism is conserved. Moreover, using computational analysis we show that antimicrobial propensity may be conserved at the N-terminus for all vertebrate RNases, thereby suggesting that a defence mechanism could be a primary function in vertebrate RNases and that the N-terminus was selected to ensure this property. In a broader context, from the overall comparison of the peptides' physicochemical and biological properties, general correlation rules could be drawn to assist in the structure-based development of antimicrobial agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Dioleoyl-sn-Glycero-3-Phosphocholin, lyophilized powder