Merck
  • One calcitriol dose transiently increases Helios+ FoxP3+ T cells and ameliorates autoimmune demyelinating disease.
One calcitriol dose transiently increases Helios+ FoxP3+ T cells and ameliorates autoimmune demyelinating disease.
Journal of neuroimmunology (2013-08-24)
Faye E Nashold, Corwin D Nelson, Lauren M Brown, Colleen E Hayes
ZUSAMMENFASSUNG
Multiple sclerosis (MS) is an incurable inflammatory demyelinating disease. We investigated one calcitriol dose plus vitamin D3 (calcitriol/+D) as a demyelinating disease treatment in experimental autoimmune encephalomyelitis (EAE). Evidence that calcitriol-vitamin D receptor pathway deficits may promote MS, and data showing calcitriol enhancement of autoimmune T cell apoptosis provided the rationale. Whereas vitamin D3 alone was ineffective, calcitriol/+D transiently increased central nervous system (CNS) Helios(+)FoxP3(+) T cells and sustainably decreased CNS T cells, pathology, and neurological deficits in mice with EAE. Calcitriol/+D, which was more effective than methylprednisolone, has potential for reversing inflammatory demyelinating disease safely and cost-effectively.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Supelco
1α,25-Dihydroxyvitamin D3 -Lösung, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Calcitriol, European Pharmacopoeia (EP) Reference Standard