Merck
  • Beneficial influence of carvedilol on urologic indices in patients with hypertension and benign prostatic hyperplasia: results of a randomized, crossover study.
Beneficial influence of carvedilol on urologic indices in patients with hypertension and benign prostatic hyperplasia: results of a randomized, crossover study.
Urology (2013-08-31)
Jacek Lewandowski, Maciej Sinski, Bartosz Symonides, Jerzy Korecki, Krzysztof Rogowski, Jacek Judycki, Artur Sieczych, Olga Możeńska, Zbigniew Gaciong
ZUSAMMENFASSUNG
To assess the influence of carvedilol, an α- and β-blocker, on lower urinary tract symptoms (LUTS) and urine flow in hypertensive patients with benign prostatic hyperplasia (BPH). Fifty men were included in this double blind crossover study with placebo. After initial screening, participants were randomized to the carvedilol or the enalapril group, with cross over after 3 months. Doses of both drugs were uptitrated or additional therapy was introduced to ensure normal control of blood pressure (BP). Urologic assessment included uroflowmetry (average [Qavg] and maximum urinary flow rate [Qmax]), postvoid residual urine volume (PVR), International Prostate Symptom Score (IPSS), and prostate-specific antigen (PSA). After carvedilol or enalapril administration, BP values were significantly reduced, whereas heart rate decreased only in the carvedilol group. Basal urologic values for carvedilol and enalapril were similar: Qavg, 7.8 ± 0.9 and 8.1 ± 0.6 mL/s; Qmax, 13.2 ± 1.5 and 13.7 ± 0.9 mL/s; PVR, 86.1 ± 13.2 and 85.6 ± 11.7 mL; and IPSS, 13.2 ± 0.9 and 12.3 ± 0.8 points, respectively. After treatment with carvedilol, PVR and IPSS significantly decreased (48.2 ± 11.7 mL, 9.0 ± 0.8 points, respectively; P <.001), whereas Qavg and Qmax increased (10.3 ± 0.9 mL/s, 16.5 ± 1.4 mL/s, respectively; P <.001). In the enalapril group, all of these values remained unchanged. Carvedilol, compared with enalapril, has a positive influence on LUTS related to BPH in patients with hypertension. Thus, therapy with carvedilol may be considered in hypertensive patients with BPH. Further studies on the urologic benefit from long-term use of the drug are warranted.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Enalapril-Maleat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Enalapril -maleat (Salz), powder, ≥98.0% (HPLC)
Sigma-Aldrich
Carvedilol, ≥98% (HPLC), solid
Supelco
Carvedilol, Pharmaceutical Secondary Standard; Certified Reference Material