Merck
  • IL-27 acts on DCs to suppress the T cell response and autoimmunity by inducing expression of the immunoregulatory molecule CD39.
IL-27 acts on DCs to suppress the T cell response and autoimmunity by inducing expression of the immunoregulatory molecule CD39.
Nature immunology (2013-09-03)
Ivan D Mascanfroni, Ada Yeste, Silvio M Vieira, Evan J Burns, Bonny Patel, Ido Sloma, Yan Wu, Lior Mayo, Rotem Ben-Hamo, Sol Efroni, Vijay K Kuchroo, Simon C Robson, Francisco J Quintana
ZUSAMMENFASSUNG
Dendritic cells (DCs) control the balance between effector T cells and regulatory T cells in vivo. Hence, the study of DCs might identify mechanisms of disease pathogenesis and guide new therapeutic approaches for disorders mediated by the immune system. We found that interleukin 27 (IL-27) signaling in mouse DCs limited the generation of effector cells of the TH1 and TH17 subsets of helper T cells and the development of experimental autoimmune encephalomyelitis (EAE). The effects of IL-27 were mediated at least in part through induction of the immunoregulatory molecule CD39 in DCs. IL-27-induced CD39 decreased the extracellular concentration of ATP and downregulated nucleotide-dependent activation of the NLRP3 inflammasome. Finally, therapeutic vaccination with IL-27-conditioned DCs suppressed established relapsing-remitting EAE. Thus, IL-27 signaling in DCs limited pathogenic T cell responses and the development of autoimmunity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Apyrase aus Kartoffeln, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase aus Kartoffeln, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase aus Kartoffeln, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase aus Kartoffeln, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder