Merck
Berberine and S allyl cysteine mediated amelioration of DEN+CCl4 induced hepatocarcinoma.
Biochimica et biophysica acta (2013-09-04)
Dipanwita Sengupta, Kaustav Dutta Chowdhury, Avik Sarkar, Soumosish Paul, Gobinda Chandra Sadhukhan
ZUSAMMENFASSUNG
Diethylnitrosamine (DEN) and carbon tetrachloride (CCl4) have been used as initiator and promoter respectively to establish an animal model for investigating molecular events appear to be involved in development of liver cancer. Use of herbal medicine in therapeutics to avoid the recurrence of hepatocarcinoma has already generated considerable interest among oncologists. In this context studies involving S-allyl-cysteine (SAC) and berberine have come up with promising results. Here we have determined the individual effect of SAC and berberine on the biomolecules associated with DEN+CCl4 induced hepatocarcinoma. Effective therapeutic value of combined treatment has also been estimated. ROS accumulation was analyzed by FACS following DCFDA incubation. Bcl2-Bax and HDAC1-pMdm2 interaction were demonstrated by co-immunoprecipitation. Immunosorbent assay was performed to analyze PP2A and caspase3 activities. MMP was determined cytofluorimetrically by investigating JC-1 fluorescence. AnnexinV binding was demonstrated by labeling the cells with AnV-FITC followed by flow cytometry. CytochromeP4502E1 mediated bioactivation of DEN+CCl4 induced Akt dependent pMdm2-HDAC1 interaction that led to p53 deacetylation, probable cause of its degradation. In parallel, oxidative stress dependent Nrf2-HO1 activation increased Bcl2 expression which in turn stimulated cell proliferation. SAC in combination with berberine inhibited Akt mediated cell proliferation. Activation of PP2A as well as inhibition of JNK resulted in induction of apoptosis after 30 days of treatment. Extension of combined treatment reverted tissue physiology towards control. Co-treated group displayed normal tissue structure. SAC and berberine mediated HDAC1/Akt inhibition implicates the efficacy of combined treatment in the amelioration of DEN+CCl4 induced hepatocarcinoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
N-Nitrosodiethylamin (NDEA) -Lösung, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
N-Nitrosodiethylamin, ≥99.0% (GC)
Sigma-Aldrich
N-Nitrosodiethylamin, liquid
Sigma-Aldrich
N-Nitrosodiethylamin, ISOPAC®
Sigma-Aldrich
S-Allyl-L-cystein, ≥98% (HPLC)
Supelco
N-Nitrosodiethylamin, analytical standard