Merck
Challenge and opportunity of targeted lung cancer chemoprevention.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-09-05)
Steven M Dubinett, Avrum Spira
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis