Merck
  • A combination of site-directed mutagenesis and chemical modification to improve diastereopreference of Pseudomonas alcaligenes lipase.
A combination of site-directed mutagenesis and chemical modification to improve diastereopreference of Pseudomonas alcaligenes lipase.
Biochimica et biophysica acta (2013-09-10)
Hui Chen, Jianping Wu, Lirong Yang, Gang Xu
ZUSAMMENFASSUNG
A combination of site-directed mutagenesis and chemical modification was employed to alter protein structure with the objective of improving diastereopreference over that achieved by simple site-directed mutagenesis. Conformational analysis using molecular dynamic (MD) simulation of Pseudomonas alcaligenes lipase (PAL) indicated that stronger steric exclusion and structural rigidity facilitated diastereopreference. A cysteine (Cys) residue was introduced using site-directed mutagenesis to construct variant A272C. The modifier 5,5'-dithiobis-(2-nitrobenzoic acid) (DTNB) was then reacted with the introduced Cys residue to provide stronger steric exclusion and structural rigidity. The modification was verified by matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) mass spectrometry. Diastereopreference was improved significantly. The diastereomeric excess (dep) of l-menthol increased from 35% with wild type PAL to 90% with A272C-DTNB modified PAL when the conversion ratio of l-menthyl propionate was nearly 100%. Conformation and kinetic parameter analysis showed that A272C-DTNB modified PAL exhibited stronger steric exclusion and increased structural rigidity around the modification site that inhibited the hydrolysis of non-targeted substrates. The combination of site-directed mutagenesis and chemical modification could be an effective method to alter protein properties and enhance diastereopreference through the combined effect of steric exclusion and structural rigidity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Menthol, 99%
Sigma-Aldrich
DL-Menthol, ≥95%, FCC, FG
Sigma-Aldrich
(±)-Menthol, racemic, ≥98.0% (GC)
Menthol, European Pharmacopoeia (EP) Reference Standard