Merck
  • Control of surface charges by radicals as a principle of antistatic polymers protecting electronic circuitry.
Control of surface charges by radicals as a principle of antistatic polymers protecting electronic circuitry.
Science (New York, N.Y.) (2013-09-21)
H Tarik Baytekin, Bilge Baytekin, Thomas M Hermans, Bartlomiej Kowalczyk, Bartosz A Grzybowski
ZUSAMMENFASSUNG
Even minute quantities of electric charge accumulating on polymer surfaces can cause shocks, explosions, and multibillion-dollar losses to electronic circuitry. This paper demonstrates that to remove static electricity, it is not at all necessary to "target" the charges themselves. Instead, the way to discharge a polymer is to remove radicals from its surface. These radicals colocalize with and stabilize the charges; when they are scavenged, the surfaces discharge rapidly. This radical-charge interplay allows for controlling static electricity by doping common polymers with small amounts of radical-scavenging molecules, including the familiar vitamin E. The effectiveness of this approach is demonstrated by rendering common polymers dust-mitigating and also by using them as coatings that prevent the failure of electronic circuitry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Siliconöl, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Siliconöl
Sigma-Aldrich
Siliconöl, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 50 cSt (25 °C)
Siliconöl, for melting point and boiling point apparatuses
Sigma-Aldrich
Siliconöl, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Siliconöl, viscosity 100,000 cSt (25 °C)