Merck
  • Expanded therapeutic potential in activity space of next-generation 5-nitroimidazole antimicrobials with broad structural diversity.
Expanded therapeutic potential in activity space of next-generation 5-nitroimidazole antimicrobials with broad structural diversity.
Proceedings of the National Academy of Sciences of the United States of America (2013-10-09)
Yukiko Miyamoto, Jaroslaw Kalisiak, Keith Korthals, Tineke Lauwaet, Dae Young Cheung, Ricardo Lozano, Eduardo R Cobo, Peter Upcroft, Jacqueline A Upcroft, Douglas E Berg, Frances D Gillin, Valery V Fokin, K Barry Sharpless, Lars Eckmann
ZUSAMMENFASSUNG
Metronidazole and other 5-nitroimidazoles (5-NI) are among the most effective antimicrobials available against many important anaerobic pathogens, but evolving resistance is threatening their long-term clinical utility. The common 5-NIs were developed decades ago, yet little 5-NI drug development has since taken place, leaving the true potential of this important drug class unexplored. Here we report on a unique approach to the modular synthesis of diversified 5-NIs for broad exploration of their antimicrobial potential. Many of the more than 650 synthesized compounds, carrying structurally diverse functional groups, have vastly improved activity against a range of microbes, including the pathogenic protozoa Giardia lamblia and Trichomonas vaginalis, and the bacterial pathogens Helicobacter pylori, Clostridium difficile, and Bacteroides fragilis. Furthermore, they can overcome different forms of drug resistance, and are active and nontoxic in animal infection models. These findings provide impetus to the development of structurally diverse, next-generation 5-NI drugs as agents in the antimicrobial armamentarium, thus ensuring their future viability as primary therapeutic agents against many clinically important infections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Nitro-imidazol, 97%