Merck
[Case of primary malignant lymphoma of the bladder].
Hinyokika kiyo. Acta urologica Japonica (2013-10-12)
Daisuke Goto, Takeshi Otani, Atsushi Tomioka, Yoshio Maruyama
ZUSAMMENFASSUNG
We report a case of primary malignant lymphoma of the bladder. An 87-year-old female visited our hospital for incidental bladder tumor. Cystoscopic examination demonstrated a non-papillary tumor over 10 mm in diameter. We performed transurethral resection of the bladder tumor. Histological examination showed malignant lymphoma, diffuse large B cell type. After further examination, it was diagnosed as primary malignant lymphoma of the bladder, stage IA E(Ann Arbor classification). We performed six courses of R-CHOP regimen (rituximab, cyclophosphamide, vincristine, doxorubicin, predonisolone). We did not find any local or distant recurrences after eight months' follow up.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vincristin -sulfat (Salz), 95.0-105.0% (HPLC), powder or crystals
Sigma-Aldrich
Vincristin -sulfat, meets USP testing specifications
Vincristinsulfat, European Pharmacopoeia (EP) Reference Standard