Merck
  • Oxaliplatin-based chemotherapy in patients aged 75 years or older with metastatic colorectal cancer.
Oxaliplatin-based chemotherapy in patients aged 75 years or older with metastatic colorectal cancer.
Anticancer research (2013-10-15)
Minoru Fukuchi, Keiichiro Ishibashi, Yusuke Tajima, Norimichi Okada, Masaru Yokoyama, Noriyasu Chika, Satoshi Hatano, Takeaki Matsuzawa, Kensuke Kumamoto, Youichi Kumagai, Hiroyuki Baba, Erito Mochiki, Hideyuki Ishida
ZUSAMMENFASSUNG
To evaluate the tolerability and efficacy of oxaliplatin-based chemotherapy (OBC) in patients ≥ 75 years old with metastatic colorectal cancer (CRC). We reviewed the medical records of 126 patients with unresectable stage IV CRC in terms of OBC administered as first-line chemotherapy whenever feasible. Use of first-line OBC was significantly less frequent in patients ≥ 75 years old (n=18) than in patients <75 years old (n=108) (46% vs. 81% p<0.01). When analysis was restricted to patients receiving OBC, the two age groups did not differ significantly in terms of response rate (44% vs. 36%, p=0.54), progression-free survival (18.7 months vs. 13.0 months, p=0.44), overall survival (25.4 months vs. 17.5 months, p=0.53), and frequency of grade 3-4 toxicity (72% vs. 58%, p=0.26). In selected patients aged 75 years or greater, the clinical outcomes of OBC seem equivalent to those of younger patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxaliplatin, powder
Oxaliplatin, European Pharmacopoeia (EP) Reference Standard