Merck
  • Primary prophylaxis of disseminated histoplasmosis in HIV patients in French Guiana: arguments for cost effectiveness.
Primary prophylaxis of disseminated histoplasmosis in HIV patients in French Guiana: arguments for cost effectiveness.
The American journal of tropical medicine and hygiene (2013-10-16)
Mathieu Nacher, Antoine Adenis, Celia Basurko, Vincent Vantilcke, Denis Blanchet, Christine Aznar, Bernard Carme, Pierre Couppié
ZUSAMMENFASSUNG
Histoplasmosis is the first cause of acquired immunodeficiency syndrome (AIDS) and AIDS-related deaths in French Guiana. Cohort data were used to determine whether primary prophylaxis with 100 mg itraconazole for patients with CD4 counts < 150/mm(3) was cost-effective with different scenarios. For a scenario where 12% of patients died, 60% were aware of their human immunodeficiency virus (HIV) infection and adherence was only 50%, primary prophylaxis would prevent 1 death and 9 cases of histoplasmosis for a cost of 36,792 Euros per averted death, 1,533 per life-year saved, 4,415 Euros per averted case, when only counting the costs of itraconazole prophylaxis. Taking into account the total costs of hospitalization showed that primary prophylaxis would allow a savings of 185,178 Euros per year. Even in a scenario of low adherence, primary prophylaxis would be cost-effective in French Guiana, and presumably in the rest of the Guianas and the Amazon.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Itraconazol, ≥98% (HPLC)