Merck
  • Loss of estrogen-related receptor α promotes hepatocarcinogenesis development via metabolic and inflammatory disturbances.
Loss of estrogen-related receptor α promotes hepatocarcinogenesis development via metabolic and inflammatory disturbances.
Proceedings of the National Academy of Sciences of the United States of America (2013-10-16)
Eui-Ju Hong, Marie-Pier Levasseur, Catherine R Dufour, Marie-Claude Perry, Vincent Giguère
ZUSAMMENFASSUNG
Estrogen-related receptor α (ERRα) is a key regulator of mitochondrial function and metabolism essential for energy-driven cellular processes in both normal and cancer cells. ERRα has also been shown to mediate bone-derived macrophage activation by proinflammatory cytokines. However, the role of ERRα in cancer in which inflammation acts as a tumor promoter has yet to be investigated. Herein we show that global loss of ERRα accelerates the development of diethylnitrosamine (DEN)-induced hepatocellular carcinoma. Biochemical and metabolomics studies revealed that loss of ERRα promotes hepatocyte necrosis over apoptosis in response to DEN due to a deficiency in energy production. We further show that increased hepatocyte death and associated compensatory proliferation observed in DEN-injured ERRα-null livers is concomitant with increased nuclear factor κB (NF-κB)-dependent transcriptional control of cytokine expression in Kupffer cells. In particular, we demonstrate that loss of ERRα-dependent regulation of the NF-κB inhibitor IκBα leads to enhanced NF-κB activity and cytokine gene activation. Our work thus shows that global loss of ERRα activity promotes hepatocellular carcinoma by independent but synergistic mechanisms in hepatocytes and Kupffer cells, implying that pharmacological manipulation of ERRα activity may have a significant clinical impact on carcinogen-induced cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
N-Nitrosodiethylamin (NDEA) -Lösung, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by gel-filtration chromatography
Sigma-Aldrich
N-Nitrosodiethylamin, ≥99.0% (GC)
Sigma-Aldrich
N-Nitrosodiethylamin, liquid
Sigma-Aldrich
N-Nitrosodiethylamin, ISOPAC®
Supelco
N-Nitrosodiethylamin, analytical standard