Merck
  • 1,2-naphthoquinone stimulates lipid peroxidation and cholesterol domain formation in model membranes.
1,2-naphthoquinone stimulates lipid peroxidation and cholesterol domain formation in model membranes.
Investigative ophthalmology & visual science (2013-10-17)
Robert F Jacob, Michael D Aleo, Yehudi Self-Medlin, Colleen M Doshna, R Preston Mason
ZUSAMMENFASSUNG
Naphthalene induces cataract formation through the accumulation of its reactive metabolite, 1,2-naphthoquinone (1,2-NQ), in the ocular lens. 1,2-NQ increases lens protein oxidation and disrupts fiber cell membrane function; however, the association of these effects with changes in membrane structure is not understood. The goal of this study was to determine the direct effects of 1,2-NQ on membrane lipid oxidation and structural organization. Iodometric approaches were used to measure the effects of naphthalene and 1,2-NQ on lipid hydroperoxide (LOOH) formation in model membranes composed of cholesterol and dilinoleoylphosphatidylcholine. Membrane samples were prepared at various cholesterol-to-phospholipid mole ratios and subjected to autoxidation at 37°C for 48 hours in the absence or presence of either agent alone (0.1-5.0 μM) or in combination with vitamin E. Small-angle x-ray diffraction was used to measure the effects of naphthalene and 1,2-NQ on membrane structure before and after exposure to oxidative stress. 1,2-NQ increased LOOH formation by 250% (P < 0.001) and 350% (P < 0.001) at 1.0 and 5.0 μM, respectively, whereas naphthalene decreased LOOH levels by 25% (P < 0.01) and 10% (NS). The pro-oxidant effect of 1,2-NQ was inversely affected by membrane cholesterol enrichment and completely blocked by vitamin E. 1,2-NQ also increased cholesterol domain formation by 360% in membranes exposed to oxidative stress; however, no significant changes in membrane lipid organization were observed with naphthalene under the same conditions. These data suggest a novel mechanism for naphthalene-induced cataract, facilitated by the direct effects of 1,2-NQ on lipid peroxidation and cholesterol domain formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Naphthalin, 99%
Supelco
Naphthalin, analytical standard
Sigma-Aldrich
Naphthalin, suitable for scintillation, ≥99%
Supelco
Schmelzpunktstandard 79-81 °C, analytical standard
Supelco
Naphthalin -Lösung, certified reference material, 5000 μg/mL in methanol
Supelco
Naphthalin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland