Merck
  • Magnolol induces apoptosis in MCF-7 human breast cancer cells through G2/M phase arrest and caspase-independent pathway.
Magnolol induces apoptosis in MCF-7 human breast cancer cells through G2/M phase arrest and caspase-independent pathway.
Die Pharmazie (2013-10-24)
Yongfeng Zhou, Yanying Bi, Chunhui Yang, Jingbo Yang, Yu Jiang, Fanmin Meng, Bo Yu, Muhammad Khan, Tonghui Ma, Hong Yang
ZUSAMMENFASSUNG
Magnolol, a small-molecule hydroxylated biphenol, isolated from the root and stem bark of Magnolia officinalis, has been shown to possess antiproliferative effect on various cancer cell lines. In the current study, we found that magnolol potently inhibited proliferation and induced apoptosis in MCF-7 human breast cancer cells. Further mechanistic studies revealed that induction of apoptosis is associated with cell cycle arrest at G2/M phase, increased generation of reactive oxygen species (ROS), reduced mitochondrial membrane potential (MMP), release of cytochrome c (Cyto c) and apoptosis inducing factor (AIF) from mitochondria to cytosol, upregulation of Bax, p21 and p53, and down-regulation of Bcl-2, cyclin B1 and cyclin-dependent kinase 1 (CDK1). Our findings indicated that magnolol induced apoptosis in MCF-7 cells via the intrinsic pathway with release of AIF from mitochondrial and G2/M phase arrest pathway. Therefore, magnolol might be a potential lead compound in the therapy of breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Cytochrom c aus Pferdeherz, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrom c aus Pferdeherz, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrom c aus Rinderherz, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrom c aus Rinderherz, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Hoechst 33258 -Lösung, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
Magnolol, ≥90% (HPLC), from plant
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
bis-Benzimid H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)
Sigma-Aldrich
bis-Benzimid H 33258, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
Cytochrom c aus Pferdeherz, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrom c aus Pferdeherz, BioReagent, suitable for GFC marker
Sigma-Aldrich
bis-Benzimid H 33258, ≥98% (HPLC and TLC)
Sigma-Aldrich
ProteoMass Cytochrom c MALDI-MS-Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Cytochrom c aus Taubenbrustmuskel, ≥95% based on Mol. Wt. 12,173 basis