Merck
  • Protective effect of lycopene on oxidative stress and antioxidant status in Cyprinus carpio during cypermethrin exposure.
Protective effect of lycopene on oxidative stress and antioxidant status in Cyprinus carpio during cypermethrin exposure.
Environmental toxicology (2013-10-24)
M Enis Yonar
ZUSAMMENFASSUNG
The aim of this study was to investigate the ameliorative properties of lycopene against the toxic effects of cypermethrin (CYP) by examining oxidative damage markers such as lipid peroxidation and the antioxidant defense system components in carp (Cyprinus carpio). The fish were divided into seven groups of 10 fish each and received the following treatments: group 1, no treatment; group 2, orally administered corn oil; group 3, oral lycopene (10 mg/kg body weight); group 4, exposure to 0.202 μg/L CYP; group 5, exposure to 0.202 μg/L CYP plus oral administration of 10 mg/kg lycopene; group 6, exposure to 0.404 μg/L CYP; and group 7, exposure to 0.404 μg/L CYP plus oral administration of 10 mg/kg lycopene. Treatment was continued for 28 days, and at the end of this period, blood and tissue (liver, kidney, and gill) samples were collected. Levels of malondialdehyde (MDA) and reduced glutathione (GSH) as well as superoxide dismutase (SOD), catalase (CAT), and glutathione peroxidase (GSH-Px) activities were determined in blood and tissues for measurement of oxidant-antioxidant status. MDA level, as an index of lipid peroxidation, increased in blood and tissues. Antioxidant enzyme activities in blood and tissues were modified in CYP groups compared with controls. Administration of lycopene ameliorated these parameters. The present results suggest that administration of lycopene might alleviate CYP-induced oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Cypermethrin, PESTANAL®, analytical standard
Supelco
α-Cypermethrin, PESTANAL®, analytical standard
Supelco
Cypermethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Lycopen, Redivivo, 10% FS, ~10% in corn oil, ≥95.0% (sum of isomers)
Supelco
Cypermethrin -Lösung, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard