- Semagacestat for treatment of Alzheimer's disease.
Semagacestat for treatment of Alzheimer's disease.
The New England journal of medicine (2013-10-25)
Rachelle S Doody, Paul S Aisen, Takeshi Iwatsubo
PMID24152267
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Alanin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Alanin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Alanin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland