Merck
Compliance with pregnancy prevention measures during isotretinoin therapy.
Journal of the American Academy of Dermatology (2013-10-26)
Mary-Katharine Collins, Jacqueline F Moreau, Dan Opel, James Swan, Noel Prevost, Matthew Hastings, Eleanor Bimla Schwarz, Laura Korb Ferris
ZUSAMMENFASSUNG
Approximately 150 women annually become pregnant while taking isotretinoin despite participation in the iPLEDGE program. Noncompliance with the requirement to be abstinent or use 2 contraceptive methods may be a contributing factor. We sought to determine the degree of adherence to contraception or abstinence among women taking isotretinoin. We conducted an anonymous survey of women of childbearing potential taking isotretinoin for at least 2 months. Among 75 participants, 21 (28%) chose abstinence as their primary means of pregnancy prevention, of whom 4 (19%) were sexually active during treatment. The most commonly chosen contraceptive methods among the 39 women who were sexually active were condoms (35, 90%) and oral contraceptive pills (18, 46%). Twelve women (31%) admitted to having intercourse at least once using 1 or fewer forms of contraception; 10 failed to use condoms, and 1 reported completely unprotected intercourse. Among sexually active oral contraceptive pill users, 7 (39%) reported missing 1 or more pills in the previous month. Data were self-reported, thus participants may have inaccurately reported contraception use. Encouraging the use of highly effective, patient-independent contraception and limiting abstinence to women who have never been sexually active may further reduce the rate of isotretinoin-exposed pregnancies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
13-cis-Retinolsäure, ≥98% (HPLC)
Supelco
Isotretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Isotretinoin, European Pharmacopoeia (EP) Reference Standard