Merck
  • [What kind of antiplatelet treatment in acute coronary syndrome?--Intensive antiplatelet treatment only for patients with invasive strategy].
[What kind of antiplatelet treatment in acute coronary syndrome?--Intensive antiplatelet treatment only for patients with invasive strategy].
Deutsche medizinische Wochenschrift (1946) (2013-10-29)
Thomas Meinertz
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Ticlopidin -hydrochlorid, analytical standard, for drug analysis
Ticlopidin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard