- Management of nocturnal enuresis.
Management of nocturnal enuresis.
BMJ (Clinical research ed.) (2013-10-31)
Patrina H Y Caldwell, Aniruddh V Deshpande, Alexander Von Gontard
PMID24170156
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Imipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Desmopressin, European Pharmacopoeia (EP) Reference Standard
Imipramin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard