Merck
  • Reduction of steroid-induced intraocular pressure elevation in sheep by tissue plasminogen activator.
Reduction of steroid-induced intraocular pressure elevation in sheep by tissue plasminogen activator.
Investigative ophthalmology & visual science (2013-11-02)
Rosana Gerometta, Sandeep Kumar, Shaily Shah, Larry Alvarez, Oscar Candia, John Danias
ZUSAMMENFASSUNG
To investigate whether tissue plasminogen activator (tPA) can prevent and/or reverse steroid-induced IOP elevation in an ovine model. Three animal groups were subjected to bilateral steroid-induced IOP elevation using thrice daily topical ocular prednisolone administration. In the first group (N = 8), one eye each of two sheep was injected intravitreally with 100 μg, 200 μg, 500 μg, or 1 mg human recombinant tPA, while contralateral eyes received vehicle. In the second group (N = 2), one eye was injected intravitreally with tPA (100 μg), while contralateral eyes received vehicle containing L-arginine. In the third group (N = 4), each animal received intravitreal tPA in one eye concurrently with initiation of bilateral steroid administration. IOP was monitored for the duration of the experiment. Tissues from eyes of the third group were used to determine relative gene expression. In the first and second groups, IOP decreased by 9.7 (±2.8) and 9.7 (±1.6) mm Hg, respectively, 24 hours after tPA administration. In the third group, tPA-treated eyes did not develop IOP elevation with ΔIOP of 11.8 (±1.3) mm Hg 8 days later. In all tPA-treated eyes, IOP remained low until the end of the study. mRNA levels in the trabecular meshwork were decreased for plasminogen activator tissue (PLAT), increased for matrix-metalloproteinase 1 (MMP-1), and stable for plasminogen activator inhibitor 1 (PAI-1), MMP-2, MMP-9, and MMP-13 in tPA-treated eyes compared with contralateral controls. PAI-1 mRNA levels in ciliary processes also remained similar. Recombinant human tPA is effective in both preventing and reversing steroid-induced IOP elevation in sheep. Tissue plasminogen activator may be useful as a therapeutic agent in steroid-induced glaucoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Prednisolon, ≥99%
Supelco
Prednisolon, Pharmaceutical Secondary Standard; Certified Reference Material
Prednisolon für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Supelco
Prednisolon, VETRANAL®, analytical standard
Prednisolon, European Pharmacopoeia (EP) Reference Standard
Prednisolon für die Systemeignung, European Pharmacopoeia (EP) Reference Standard