Merck
  • Effects of peroxisome proliferator-activated receptor-γ activation on apoptosis in rats with acute pancreatitis.
Effects of peroxisome proliferator-activated receptor-γ activation on apoptosis in rats with acute pancreatitis.
Digestive diseases and sciences (2013-11-05)
Ping Xu, Xiao-Li Lou, Cheng Chen, Zhi-Wen Yang
ZUSAMMENFASSUNG
To investigate the effects and mechanisms of peroxisome proliferator-activated receptor-γ (PPAR-γ) activation on the induction of apoptosis in rats with acute pancreatitis. Severe acute pancreatitis (SAP) and mild acute pancreatitis (MAP) were induced and pre-treated with pioglitazone, which is a ligand of PPAR-γ. The expression of inflammatory factors (TNF-α and IL6) of the pancreas was detected by ELISA. The apoptosis in pancreas were detected by TUNEL assay and the activity of caspase 3 was determined. Phosphorylation of p65 in pancreas of SAP or MAP was determined by western-blot. Expression levels of PPAR-γ proteins were elevated in the pancreases of SAP or MAP rats pre-injected with pioglitazone intraperitoneally. Downregulation of the expression TNF-α and IL6 and relief of pathological changes in the pancreas suggested that pioglitazone had protective effects on acute panceatitis. In pioglitazone pre-treated groups, a TUNEL assay indicated a high level of apoptosis in SAP but little apoptosis in MAP, showing pioglitazone could promote taurocholate-induced apoptosis but inhibit ceruleininduced apoptosis in pancraeatic aniniar cells. Furthermore, caspase 3 activity was high in SAP but low in MAP, implying that the apoptotic mechanism in pancreatic acinar cells of AP rats was correlated with caspase 3 activity. Phosphorylation of p65 was reduced in SAP or MAP group pretreated with pioglitazone, indicating that pioglitazone reduced the inflammation reaction by inhibiting the activation of the NF-κB. These results indicated that activation of PPAR-γ induced apoptosis in pancreatic acinar cells of SAP rats but inhibited apoptosis in pancraeatic acinar cells of MAP rats, which demonstrated that PPAR-γ may be an efficiently therapeutic target in pancreatic inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pioglitazon -hydrochlorid, ≥98% (HPLC)