Merck
  • Negative gate bias and light illumination-induced hump in amorphous InGaZnO thin film transistor.
Negative gate bias and light illumination-induced hump in amorphous InGaZnO thin film transistor.
Journal of nanoscience and nanotechnology (2013-11-20)
Jae-Hong Jeon, Seung-Bum Seo, Han-Sung Park, Hee-Hwan Choe, Jong-Hyun Seo, Kee-Chan Park, Sang-Hee Ko Park
ZUSAMMENFASSUNG
While observing the transfer characteristics of a-IGZO TFTs, it was noticed that a hump occurred in the subthreshold regime after light and bias stress. This study analyzes the mechanism of the hump occurrence. It was determined that hump characteristics were related with parasitic TFTs which formed at the peripheral edges parallel with the channel direction. It seems that the negative shift of the transfer characteristics of parasitic TFTs was larger than that of the main TFT under light and bias stress. Therefore, the difference in the negative shift between the main TFT and the parasitic TFT was the origin of the hump occurrence. We investigated the instability of a-IGZO TFTs under negative gate bias with light illumination for various channel structures in order to verify the above mechanism.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinkoxid, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Zinkoxid, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinkoxid, nanopowder, ≤50 nm particle size
Sigma-Aldrich
Zinkoxid, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Gallium, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Gallium(III)-oxid, ≥99.99% trace metals basis
Sigma-Aldrich
Zinkoxid, Dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinkoxid, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Zinkoxid, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium(III)-oxid, 99.99% trace metals basis
Sigma-Aldrich
Gallium, 99.9995% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinkoxid, 99.999% trace metals basis
Sigma-Aldrich
Indium(III)-oxid, 99.998% trace metals basis
Sigma-Aldrich
Gallium, 99.999% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium(III)-oxid, nanopowder, <100 nm particle size (TEM), 99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Supelco
Zinkoxid, analytical standard
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, bars, 99.95%