Merck
  • A simple preparation of a stable CdS-polyacrylamide nanocomposite: structure, thermal and optical properties.
A simple preparation of a stable CdS-polyacrylamide nanocomposite: structure, thermal and optical properties.
Journal of nanoscience and nanotechnology (2013-11-20)
Long Giang Bach, M Rafiqul Islam, Seong-Soo Hong, Kwon Taek Lim
ZUSAMMENFASSUNG
A facile approach was employed for the preparation of stable luminescent nanocomposites of CdS quantum dots (QDs) and polyacrylamide (PAM) through the cross coupling chemistry. Initially, CdS QDs were synthesized in a well controlled manner using 3-mercaptopropionic acid as a capping agent. Then, carboxylic acid groups on CdS QDs were chemically incorporated into PAM matrices with the assistance of 1-ethyl-3-(3-dimethylaminopropyl)-carbodiimide hydrochloride and N-hydroxysulfosuccinimide coupling agents. FT-IR analysis was used to investigate the chemical incorporation of CdS QDs in PAM matrices via the covalent protocol. The XPS elemental mapping studies further suggested the formation of CdS-PAM nanocomposites. FE-SEM and TEM images were engaged to study the morphologies, and distribution of CdS QDs in the PAM networks. The physical structure of the CdS-PAM nanocomposites was investigated by XRD analysis. Thermal stability of the nanocomposites was observed to be enhanced in compare to PAM as evidenced from TGA data. The UV-vis and photoluminescence studies of the CdS-PAM nanocomposites suggested their promising optical applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cadmiumselenid, -325 Mesh particle size, 99.99% trace metals basis, electronic grade