Merck
[Granulomatous sporotrichosis: report of two unusual cases].
Revista chilena de infectologia : organo oficial de la Sociedad Chilena de Infectologia (2013-11-20)
Max Ramírez-Soto, José Lizárraga-Trujillo
ZUSAMMENFASSUNG
Sporotrichosis is a subcutaneous mycosis caused by Sporothrix complex, endemic in Abancay, Peru. Is acquired by traumatic inoculation with plant material. Common clinical presentations are lymphatic cutaneous and fixed cutaneous disease. We report 2 cases of fixed cutaneous sporotrichosis with granulomatous appearance. The first case was a patient of 65 years old with no risk factors and the second case was a 67 year old diabetic patient. Subjects underwent mycological culture with Sabouraud agar, with isolation of Sporothrix schenckii and clinical dignosis of fixed cutaneous sporotrichosis with granulomatous appearance. One patient received oral treatment with saturated solution of potassium iodide (SSKI) with a initial dose of 3 drops tid up to a maximum dose of 40 drops tid. Mycological and clinical cure was achieved after 2 months of treatment. We should consider the unusual clinical presentations of fixed cutaneous sporotrichosis with granulomatous appearance that present morphological and clinical features in diabetic and nondiabetic patients older than 60 years from endemic areas and communicate adequate response to treatment with SSKI in one case.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumiodid, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumiodid, ReagentPlus®, 99%
Sigma-Aldrich
Kaliumiodid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Goldätzmittel, Standard
Sigma-Aldrich
Kaliumiodid, ≥99.99% trace metals basis
Sigma-Aldrich
Kaliumiodid, suitable for plant cell culture
Sigma-Aldrich
Kaliumiodid, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Kaliumiodid, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumiodid, AnhydroBeads, −10 mesh, 99.998% trace metals basis