Merck
Diagnosis and management of hyperhidrosis.
BMJ (Clinical research ed.) (2013-11-28)
R A Benson, R Palin, P J E Holt, I M Loftus
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aluminiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Aluminiumchlorid, reagent grade, 98%
Sigma-Aldrich
Aluminiumchlorid, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminiumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Aluminiumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminiumchlorid -Lösung, 1.0 M in nitrobenzene