Merck
Two new phthalides with BuChE inhibitory activity from Ligusticum chuanxiong.
Journal of Asian natural products research (2013-11-28)
Jian Huang, Xiao-Qing Lu, Jin Lu, Guo-Yu Li, Hang-Yu Wang, Li-Hong Li, Rui-Chao Lin, Jin-Hui Wang
ZUSAMMENFASSUNG
Two new phthalides, chuanxiongdiolides A and B, were isolated from the roots of Ligusticum chuanxiong Hort. Their structures were established by UV, IR, 1D (¹H, ¹³C) and 2D (HSQC, ¹H-¹H COSY, HMBC, NOESY) NMR, and HR-ESI-MS methods, and their absolute configurations were assigned via circular dichroism exciton chirality. The two compounds showed different degrees of inhibitory effects against butyrylcholine esterase.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phthalid, 98%