Merck
The role of vitamin D in depression: from a curious idea to a therapeutic option.
The Journal of clinical psychiatry (2013-12-18)
Cédric Annweiler, Reza Rastmanesh, Stéphane Richard-Devantoy, Olivier Beauchet
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
25-Hydroxyvitamin D2, ≥95.0% (HPLC)
Supelco
25-Hydroxyvitamin D2 -Lösung, 50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®