Merck
  • A multidisciplinary approach to study the effects of balneotherapy and mud-bath therapy treatments on fibromyalgia.
A multidisciplinary approach to study the effects of balneotherapy and mud-bath therapy treatments on fibromyalgia.
Clinical and experimental rheumatology (2014-01-01)
Laura Bazzichi, Ylenia Da Valle, Alessandra Rossi, Camillo Giacomelli, Francesca Sernissi, Gino Giannaccini, Laura Betti, Federica Ciregia, Laura Giusti, Pietro Scarpellini, Liliana Dell'Osso, Donatella Marazziti, Stefano Bombardieri, Antonio Lucacchini
ZUSAMMENFASSUNG
To study the effects of both balneotherapy and mud-bath therapy treatments in patients affected by primary fibromyalgia (FM) using rheumatological, psychiatric, biochemical and proteomic approaches. Forty-one FM patients (39 females, 2 males), who fulfilled the American College of Rheumatology criteria received a 2-week thermal therapy programme consisting of therapy once daily for 6 days/week. Twenty-one patients received mud-bath treatment, while the other twenty balneotherapy. Pain, symptoms, and quality of life were assessed. Oxytocin, brain-derived neurotrophic factor (BDNF), ATP and serotonin transporter levels during therapy were assayed. Comparative whole saliva (WS) proteomic analysis was performed using a combination of two-dimensional electrophoresis (2DE) and mass spectrometry techniques. We observed a reduction in pain, FIQ values and improvement of SF36 in both groups of patients treated with mud-bath or balneotherapy. The improvement of the outcome measures occurred with different timing and duration in the two spa treatments. A significant decrease in BDNF concentrations was observed either after balneotherapy or mud-bath therapy when assayed after twelve weeks, while no significant change in oxytocin levels, ATP levels and serotonin transporter were detected. Significant differences were observed for phosphoglycerate mutase1 (PGAM1) and zinc alpha-2-glycoprotein 1 (AZGP1) protein expression. Our results showed that the thermal treatment might have a beneficial effect on the specific symptoms of the disease. In particular, while balneotherapy gives results that in most patients occur after the end of the treatment but which are no longer noticeable after 3 months, the mud-bath treatment gives longer lasting results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oxytocin
Sigma-Aldrich
Oxytocin, lyophilized powder, ~15 IU/mg solid (Prepared from synthetic oxytocin)
Sigma-Aldrich
Oxytocin acetate salt hydrate, ≥97% (HPLC)
Oxytocin, European Pharmacopoeia (EP) Reference Standard