Merck
Role for terminal complement activation in amyotrophic lateral sclerosis disease progression.
Proceedings of the National Academy of Sciences of the United States of America (2014-01-02)
Trent M Woodruff, John D Lee, Peter G Noakes
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Komplementkomponente C1q aus Humanserum, ≥95% (SDS-PAGE)