Merck
  • Secondary oxalosis due to excess vitamin C intake: a cause of graft loss in a renal transplant recipient.
Secondary oxalosis due to excess vitamin C intake: a cause of graft loss in a renal transplant recipient.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia (2014-01-18)
S Yaich, Y Chaabouni, K Charfeddine, S Zaghdane, M Kharrat, K Kammoun, S Makni, T Boudawara, J Hachicha
ZUSAMMENFASSUNG
Renal oxalate deposition can be seen with primary hyperoxaluria, malabsorptive states, ethylene glycol toxicity and, rarely, with excessive vitamin C ingestion. We report a case of secondary hyperoxaluria in which the diagnosis was not considered initially because there was no past history of urinary calculi and no evidence of nephrocalcinosis on plain X-ray of the abdomen and ultrasonography. The disease was detected and diagnosed only after kidney transplantation. Secondary oxalosis can cause graft loss or delayed graft function. Biopsy of the allograft should be carefully examined for oxalate deposits even in the absence of a family history. When oxalosis is diagnosed, intensifying hemodialysis (HD) to eliminate calcium oxalate can help in the recovery of renal function in some cases. Systematic vitamin C supplementation in HD patients should be avoided as it can be a cause of secondary oxalosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
(+)-Natrium-L-ascorbat, crystalline, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Supelco
Ascorbinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)-Natrium-L-ascorbat, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Natrium-L-ascorbat, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbinsäure, FCC, FG
Supelco
Natriumascorbat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbinsäure, BioUltra, ≥99.5% (RT)
Ascorbinsäure, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ≥99.0% (RT)
Supelco
L-Ascorbinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbinsäure, tested according to Ph. Eur.