Merck
  • [Sodium-losing nephropathy caused by tacrolimus after allogeneic hematopoietic stem cell transplantation].
[Sodium-losing nephropathy caused by tacrolimus after allogeneic hematopoietic stem cell transplantation].
[Rinsho ketsueki] The Japanese journal of clinical hematology (2014-01-24)
Sayako Yuda, Takehiko Mori, Jun Kato, Yuya Koda, Sumiko Kohashi, Taku Kikuchi, Masuho Saburi, Shinichiro Okamoto
ZUSAMMENFASSUNG
A 39-year-old woman (Case 1) and a 57-year-old woman (Case 2) underwent allogeneic bone marrow transplantation for acute lymphoblastic leukemia and follicular lymphoma, respectively. Both patients had received tacrolimus orally for treatment of or prophylaxis against graft-versus-host disease. Seventeen months (Case 1) and 2 months (Case 2) post-transplantation, when the trough level of tacrolimus was maintained around 10 ng/ml, the serum sodium levels of Cases 1 and 2 decreased to 123.5 mEq/l and 125.6 mEq/l, respectively. Urinary sodium excretions increased to 186.8 mEq/day and 375.7 mEq/day, respectively. Sodium-losing nephropathy due to tacrolimus was diagnosed, and reducing the dose of tacrolimus with no other intervention resulted in resolution of the hyponatremia. Although sporadic kidney transplantation cases with sodium-losing nephropathy due to tacrolimus have been reported, no prior cases with this complication after hematopoietic stem cell transplantation (HSCT) have been reported. Sodium-losing nephropathy should be recognized as one of the renal toxicities of tacrolimus in HSCT as well as kidney recipients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natrium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Natrium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
Natrium, 99.95% trace metals basis, ingot
Sigma-Aldrich
FK-506 Monohydrat, ≥98% (HPLC)
Sigma-Aldrich
Natrium, ACS reagent, dry
Sigma-Aldrich
Natrium, 25-35 wt % dispersion in paraffin