Merck
  • Human SAP18 mediates assembly of a splicing regulatory multiprotein complex via its ubiquitin-like fold.
Human SAP18 mediates assembly of a splicing regulatory multiprotein complex via its ubiquitin-like fold.
RNA (New York, N.Y.) (2010-10-23)
Kusum K Singh, Steffen Erkelenz, Stephanie Rattay, Anna Katharina Dehof, Andreas Hildebrandt, Klaus Schulze-Osthoff, Heiner Schaal, Christian Schwerk
ZUSAMMENFASSUNG
RNPS1, Acinus, and SAP18 form the apoptosis- and splicing-associated protein (ASAP) complex, which is also part of the exon junction complex. Whereas RNPS1 was originally identified as a general activator of mRNA processing, all three proteins have been found within functional spliceosomes. Both RNPS1 and Acinus contain typical motifs of splicing regulatory proteins including arginine/serine-rich domains. Due to the absence of such structural features, however, a function of SAP18 in splicing regulation is completely unknown. Here we have investigated splicing regulatory activities of the ASAP components. Whereas a full-length Acinus isoform displayed only limited splicing regulatory activity, both RNPS1 and, surprisingly, SAP18 strongly modulated splicing regulation. Detailed mutational analysis and three-dimensional modeling data revealed that the ubiquitin-like fold of SAP18 was required for efficient splicing regulatory activity. Coimmunoprecipitation and immunofluorescence experiments demonstrated that SAP18 assembles a nuclear speckle-localized splicing regulatory multiprotein complex including RNPS1 and Acinus via its ubiquitin-like fold. Our results therefore suggest a novel function of SAP18 in splicing regulation.