- Mammalian Trit1 is a tRNA([Ser]Sec)-isopentenyl transferase required for full selenoprotein expression.
Mammalian Trit1 is a tRNA([Ser]Sec)-isopentenyl transferase required for full selenoprotein expression.
The Biochemical journal (2013-01-08)
Noelia Fradejas, Bradley A Carlson, Eddy Rijntjes, Niels-Peter Becker, Ryuta Tobe, Ulrich Schweizer
PMID23289710
ZUSAMMENFASSUNG
Selenoproteins are proteins carrying the rare amino acid Sec (selenocysteine). Full expression of selenoproteins requires modification of tRNA([Ser]Sec), including N(6)-isopentenylation of base A(37). We show that Trit1 is a dimethylallyl:tRNA([Ser]Sec) transferase. Knockdown of Trit1 reduces expression of selenoproteins. Incubation of in vitro transcribed tRNA[Ser]Sec with recombinant Trit1 transfers [(14)C]dimethylallyl pyrophosphate to tRNA([Ser]Sec). 37A>G tRNA([Ser]Sec) is resistant to isopentenylation by Trit1.