Merck
  • 591. Amine oxidation. Part I. The side-chain oxidation of N-alkyl-and NN-dialkyl-anilines by manganese dioxide.
591. Amine oxidation. Part I. The side-chain oxidation of N-alkyl-and NN-dialkyl-anilines by manganese dioxide.
Journal of the Chemical Society (1957)
Henbest HB and Thomas A.