Merck
A RAS recruitment screen identifies ZKSCAN4 as a glucocorticoid receptor-interacting protein.
Journal of molecular endocrinology (2008-11-15)
Karin Ecker, Andreas Lorenz, Frank Wolf, Christian Ploner, Günther Böck, Tod Duncan, Stephan Geley, Arno Helmberg
ZUSAMMENFASSUNG
To search for proteins interacting with the glucocorticoid receptor, we adapted Aronheim's reverse RAS recruitment system relying on the Saccharomyces cerevisiae mutant cdc25-2, which has a temperature-dependent defect in its RAS signaling pathway driving proliferation. The full-length human glucocorticoid receptor (NR3C1, isoform-alpha) was attached to the yeast plasma membrane in either of two orientations and used as bait to screen a HeLa cell cDNA library. Library proteins were fused to constitutively active, soluble human RAS, complementing the defective yeast pathway in case of bait-prey interaction. Screening of 800 000 clones resulted in the isolation of 21 proteins, 8 of which were followed up to evaluate interaction with the receptor in human cell lines. One of these candidates, the SCAN- and KRAB-domain-containing zinc finger protein 307 (ZKSCAN4) was co-precipitated with the receptor when both proteins were overexpressed in HEK293 cells. Rabbit antisera against ZKSCAN4 were raised, affinity purified, and used to immunoprecipitate endogenous ZKSCAN4 from Hct116 cells, resulting in co-precipitation of endogenous glucocorticoid receptor. Overexpressed ZKSCAN4 was found to co-localize in granular nuclear structures with the activated glucocorticoid receptor and partially with chromatin regions characterized by histone H3 mono-methylated on lysine 4 (H3K4me1). Overexpressed ZKSCAN4 had no effect on an episomal glucocorticoid receptor-driven reporter plasmid. By contrast, ZKSCAN4 markedly reduced glucocorticoid induction of the mouse mammary tumor virus-promoter-driven reporter gene when this was chromosomally integrated, arguing for a chromatin-dependent inhibition of glucocorticoid receptor-mediated transactivation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)