Merck
  • Identification of a novel human subfamily of mitochondrial carriers with calcium-binding domains.
Identification of a novel human subfamily of mitochondrial carriers with calcium-binding domains.
The Journal of biological chemistry (2004-04-01)
Araceli del Arco, Jorgina Satrústegui
ZUSAMMENFASSUNG
Aralar1 and citrin were identified as calcium binding aspartate/glutamate carriers (AGC) in mitochondria. The presence of calcium binding motifs facing the extramitochondrial space allows the regulation of the transport activity of these carriers by cytosolic calcium and provides a new mechanism to transduce calcium signals in mitochondria without the requirement of calcium entry in the organelle. We now report the complete characterization of a second subfamily of human calcium binding mitochondrial carriers named SCaMC (short calcium-binding mitochondrial carriers). We have identified three SCaMC genes in the human genome. All code for highly conserved proteins (about 70-80% identity), of about 500 amino acids with a characteristic mitochondrial carrier domain at the C terminus, and an N-terminal extension harboring four EF-hand binding motifs with high similarity to calmodulin. All SCaMC proteins were found to be located exclusively in mitochondria, and their N-terminal extensions were dispensable for the correct mitochondrial targeting of the polypeptides. SCaMC-1 is the human orthologue of the rabbit Efinal protein, which was reported to be located in peroxisomes, and SCaMC-2 is the human orthologue of the rat MCSC protein, described as up-regulated by dexamethasone in AR42J cells. One of the SCaMC genes, SCaMC-2, has four variants generated by alternative splicing, resulting in proteins with a common C terminus but with variations in their N-terminal halves, including the loss of one to three EF-hand motifs. These results make SCaMC one of most complex subfamilies of mitochondrial carriers and suggest that the large number of isoforms and splice variants may confer different calcium sensitivity to the transport activity of these carriers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SLC25A24 antibody produced in rabbit, IgG fraction of antiserum