Merck
  • Room-temperature phosphorescence from 1: 1: 1 inclusion compounds of. beta.-cyclodextrin with brominated alcohols and acenaphthene.
Room-temperature phosphorescence from 1: 1: 1 inclusion compounds of. beta.-cyclodextrin with brominated alcohols and acenaphthene.
Journal of the American Chemical Society (1989)
Hamai S.